Assay Details
| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045008.1 | - | 8 - 9 | 1212 | 75 | |
| U16162.1 | - | 7 - 8 | 1053 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P4ha1| Gene Symbol: P4ha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045008.1 | - | 8 - 9 | 1212 | 75 | |
| U16162.1 | - | 7 - 8 | 1053 | 75 | ||
Set Membership: |
|