Assay Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030253.3 | NP_084529.1 | 1 - 2 | 592 | 68 | |
| GenBank mRNA | AK037903.1 | - | 1 - 2 | 51 | 68 | |
| AK050032.1 | - | 1 - 2 | 36 | 68 | ||
| BC010312.1 | - | 1 - 2 | 51 | 68 | ||
| BC070466.1 | - | 1 - 2 | 592 | 30615225 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037903.1, AK050032.1, BC070466.1, NM_030253.3 transcripts.
Set Membership: |
|

Parp9
Genomic Map