Assay Details
| Gene Symbol: Aqp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073483.1 | 4 - 5 | 704 | 93 | ||
| GenBank mRNA | BC024105.1 | - | 4 - 5 | 675 | 5096952 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Aqp9
Genomic Map