Assay Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008246.1 | - | 1 - 2 | 332 | 63 | |
| AK008317.1 | - | 1 - 2 | 333 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008246.1, AK008317.1 transcripts.
Set Membership: |
|

Dpagt1
Genomic Map