Assay Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023635.6 | NP_076124.1 | 1 - 2 | 202 | 92 | |
| GenBank mRNA | AK010373.1 | - | 1 - 2 | 120 | 92 | |
| AK030529.1 | - | 1 - 2 | 149 | 92 | ||
| BC009656.1 | - | 1 - 2 | 155 | 3590089 | 92 | |
| CJ077071.1 | - | 1 - 2 | 202 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010373.1, AK030529.1, BC009656.1, NM_023635.6 transcripts.
Set Membership: |
|

Rab27a
Genomic Map