Assay Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110300.1 | NP_001103770.1 | 5 - 6 | 757 | 66 | |
| GenBank mRNA | AK010478.1 | - | 5 - 6 | 736 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1m2
Genomic Map