Assay Details
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110307.1 | NP_001103777.1 | 1 - 2 | 260 | 74 | |
GenBank mRNA | AK004738.1 | - | 1 - 2 | 260 | 74 | |
AK150437.1 | - | 1 - 2 | 164 | 74 | ||
AK152142.1 | - | 1 - 2 | 164 | 74 | ||
AK154430.1 | - | 1 - 2 | 75 | 74 | ||
AK155507.1 | - | 1 - 2 | 75 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK004738.1, AK150437.1, AK152142.1, AK154430.1, AK155507.1, NM_001110307.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
