Assay Details
| Gene Symbol: Ccdc82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025534.2 | NP_079810.2 | 6 - 7 | 1274 | 104 | |
| GenBank mRNA | BC040405.1 | - | 4 - 5 | 435 | 104 | |
| BC057326.1 | - | 6 - 7 | 1274 | 6826080 | 104 | |
| BC098496.1 | - | 5 - 6 | 1201 | 30089818 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc82
Genomic Map