Assay Details
| Gene Symbol: Matk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285855.1 | NP_001272784.1 | 1 - 2 | 342 | 69 | |
| GenBank mRNA | D45243.1 | - | 1 - 2 | 342 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D45243.1, NM_001285855.1 transcripts.
Set Membership: |
|

Matk
Genomic Map