Assay Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145611.4 | NP_663586.3 | 7 - 8 | 1970 | 60 | |
| GenBank mRNA | AK031324.1 | - | 7 - 8 | 1963 | 60 | |
| AK045492.1 | - | 7 - 8 | 1942 | 60 | ||
| AK049288.1 | - | 7 - 8 | 1971 | 60 | ||
| BC006647.1 | - | 7 - 8 | 1958 | 60 | ||
More Information
Set Membership: |
|

Kank2
Genomic Map