Assay Details
| Gene Symbol: Nrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032002.2 | NP_114391.1 | 3 - 4 | 289 | 98 | |
| GenBank mRNA | AF083067.2 | - | 2 - 3 | 167 | 98 | |
| AK080089.1 | - | 3 - 4 | 267 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrg4
Genomic Map