Assay Details
| Gene Symbol: Nrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032002.2 | NP_114391.1 | 4 - 5 | 437 | 67 | |
| GenBank mRNA | AF083067.2 | - | 3 - 4 | 315 | 67 | |
| AK041077.1 | - | 5 - 6 | 469 | 67 | ||
| AK080058.1 | - | 4 - 5 | 389 | 67 | ||
| BC034839.1 | - | 7 - 8 | 730 | 3372942 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrg4
Genomic Map