Assay Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013852.2 | NP_038880.1 | 10 - 11 | 1106 | 78 | |
| GenBank mRNA | AK030626.1 | - | 10 - 11 | 1071 | 78 | |
| AK177336.1 | - | 2 - 3 | 102 | 78 | ||
| AK179781.1 | - | 4 - 5 | 336 | 78 | ||
| AK211939.1 | - | 2 - 3 | 102 | 78 | ||
| BC032923.1 | - | 10 - 11 | 1047 | 1430389 | 78 | |
More Information
Set Membership: |
|

Abcf3
Genomic Map