| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600002644.mm.1 |
NM_013852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002644.mm.1 |
NM_013852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002644.mm.1 |
TC1600000280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600002640.mm.1 |
NM_013852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002640.mm.1 |
NM_013852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002640.mm.1 |
TC1600000280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600002645.mm.1 |
NM_013852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002645.mm.1 |
NM_013852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002645.mm.1 |
TC1600000280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600002642.mm.1 |
NM_013852.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002642.mm.1 |
NM_013852.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600002642.mm.1 |
TC1600000280.mm.1 |
MTA TC |