Assay Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013852.2 | NP_038880.1 | 3 - 4 | 370 | 67 | |
| GenBank mRNA | AK030626.1 | - | 3 - 4 | 334 | 67 | |
| BC032923.1 | - | 3 - 4 | 311 | 1430389 | 67 | |
| CD540120.1 | - | 3 - 4 | 370 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcf3
Genomic Map