Assay Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013852.2 | NP_038880.1 | 4 - 5 | 420 | 75 | |
| GenBank mRNA | AK030626.1 | - | 4 - 5 | 384 | 75 | |
| AK214630.1 | - | 1 - 2 | 51 | 75 | ||
| BC032923.1 | - | 4 - 5 | 361 | 1430389 | 75 | |
| CD540120.1 | - | 4 - 5 | 420 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcf3
Genomic Map