Assay Details
| Gene Symbol: Abcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013852.2 | NP_038880.1 | 5 - 6 | 514 | 75 | |
| GenBank mRNA | AK030626.1 | - | 5 - 6 | 478 | 75 | |
| AK183731.1 | - | 3 - 4 | 216 | 75 | ||
| AK214630.1 | - | 2 - 3 | 145 | 75 | ||
| BC032923.1 | - | 5 - 6 | 455 | 1430389 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcf3
Genomic Map