Assay Details
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207676.2 | NP_997559.1 | 7 - 8 | 1023 | 76 | |
| GenBank mRNA | AB183400.1 | - | 7 - 8 | 1002 | 76 | |
| BC057125.1 | - | 7 - 8 | 1011 | 6414884 | 76 | |
| BC090970.1 | - | 7 - 8 | 1202 | 6703838 | 76 | |
| BC094661.1 | - | 4 - 5 | 573 | 76 | ||
| DQ279856.1 | - | 7 - 8 | 1002 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cadm1
Genomic Map