Assay Details
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007713.4 | NP_031739.3 | 11 - 12 | 1999 | 85 | |
| GenBank mRNA | AF033565.1 | - | 11 - 12 | 1275 | 85 | |
| AK009908.1 | - | 7 - 8 | 718 | 85 | ||
| AK090215.1 | - | 11 - 12 | 1240 | 85 | ||
| AK133936.1 | - | 11 - 12 | 1245 | 85 | ||
| AK136481.1 | - | 9 - 10 | 3638 | 85 | ||
| AK145880.1 | - | 11 - 12 | 1254 | 85 | ||
| AK146942.1 | - | 11 - 12 | 2002 | 85 | ||
| AK154003.1 | - | 11 - 12 | 1248 | 85 | ||
| AK167300.1 | - | 11 - 12 | 2001 | 85 | ||
| AK184905.1 | - | 3 - 4 | 191 | 85 | ||
| AK190424.1 | - | 3 - 4 | 192 | 85 | ||
| AK208152.1 | - | 3 - 4 | 192 | 85 | ||
| BC085084.1 | - | 11 - 12 | 1242 | 6309020 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clk3
Genomic Map