Assay Details
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007713.4 | NP_031739.3 | 8 - 9 | 1700 | 98 | |
| GenBank mRNA | AF033565.1 | - | 8 - 9 | 976 | 98 | |
| AK009908.1 | - | 4 - 5 | 419 | 98 | ||
| AK090215.1 | - | 8 - 9 | 941 | 98 | ||
| AK133936.1 | - | 8 - 9 | 946 | 98 | ||
| AK136481.1 | - | 6 - 7 | 3339 | 98 | ||
| AK140015.1 | - | 7 - 8 | 3043 | 98 | ||
| AK145880.1 | - | 8 - 9 | 955 | 98 | ||
| AK146942.1 | - | 8 - 9 | 1703 | 98 | ||
| AK154003.1 | - | 8 - 9 | 949 | 98 | ||
| AK167300.1 | - | 8 - 9 | 1702 | 98 | ||
| AK185120.1 | - | 3 - 4 | 321 | 98 | ||
| BC085084.1 | - | 8 - 9 | 943 | 6309020 | 98 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clk3
Genomic Map