Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900015321.mm.1 |
NM_001110309.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015321.mm.1 |
NM_001110309.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015321.mm.1 |
NM_146221.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015321.mm.1 |
NM_146221.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015321.mm.1 |
TC0900001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015323.mm.1 |
NM_001110309.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015323.mm.1 |
NM_001110309.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015323.mm.1 |
NM_146221.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015323.mm.1 |
NM_146221.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015323.mm.1 |
TC0900001836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900015320.mm.1 |
NM_001110309.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015320.mm.1 |
NM_001110309.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015320.mm.1 |
NM_146221.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015320.mm.1 |
NM_146221.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900015320.mm.1 |
TC0900001836.mm.1 |
MTA TC |