Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900014617.mm.1 |
NM_027823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014617.mm.1 |
NM_027823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014617.mm.1 |
TC0900001732.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014619.mm.1 |
NM_027823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014619.mm.1 |
NM_027823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014619.mm.1 |
TC0900001732.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014621.mm.1 |
NM_027823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014621.mm.1 |
NM_027823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014621.mm.1 |
TC0900001732.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014620.mm.1 |
NM_027823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014620.mm.1 |
NM_027823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014620.mm.1 |
TC0900001732.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900014618.mm.1 |
NM_027823.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014618.mm.1 |
NM_027823.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900014618.mm.1 |
TC0900001732.mm.1 |
MTA TC |