Assay Details
Gene Symbol: Mb21d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125721.1 | 2 - 3 | 989 | 95 | ||
GenBank mRNA | BC044854.1 | - | 2 - 3 | 649 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
