Assay Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175549.4 | NP_780758.3 | 11 - 12 | 1678 | 80 | |
| GenBank mRNA | AK035110.1 | - | 12 - 13 | 2564 | 80 | |
| AK085461.1 | - | 12 - 13 | 2315 | 80 | ||
| BC055333.1 | - | 12 - 13 | 1998 | 6409283 | 80 | |
| DQ533875.1 | - | 13 - 14 | 1970 | 80 | ||
| DQ533876.1 | - | 11 - 12 | 1678 | 80 | ||
More Information
Set Membership: |
|

Robo2
Genomic Map