Assay Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175549.4 | NP_780758.3 | 15 - 16 | 2333 | 92 | |
| GenBank mRNA | AK035110.1 | - | 16 - 17 | 3218 | 92 | |
| AK085461.1 | - | 16 - 17 | 2969 | 92 | ||
| AK129396.1 | - | 2 - 3 | 312 | 92 | ||
| BC055333.1 | - | 16 | 2653 | 6409283 | 92 | |
| DQ533875.1 | - | 17 - 18 | 2625 | 92 | ||
| DQ533876.1 | - | 15 - 16 | 2333 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Robo2
Genomic Map