Assay Details
| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033359.2 | NP_001028531.2 | 13 - 14 | 2002 | 66 | |
| NM_001170787.1 | NP_001164258.1 | 17 - 18 | 2666 | 66 | ||
| GenBank mRNA | AK138164.1 | - | 13 - 14 | 2003 | 66 | |
| AK162856.1 | - | 13 - 14 | 2002 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn5
Genomic Map