Assay Details
| Gene Symbol: Pgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163746.1 | NP_001157218.1 | 10 - 11 | 1353 | 69 | |
| GenBank mRNA | AK049758.1 | - | 10 - 11 | 1340 | 69 | |
More Information
Set Membership: |
|

Pgm3
Genomic Map