Assay Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019797.4 | NP_062771.2 | 3 - 4 | 744 | 89 | |
| GenBank mRNA | AF539614.1 | - | 3 - 4 | 627 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF539614.1, NM_019797.4 transcripts.
Set Membership: |
|

Trip4
Genomic Map