Assay Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008988.2 | NP_033014.1 | 11 - 12 | 2117 | 76 | |
| GenBank mRNA | AF026465.1 | - | 11 - 12 | 2117 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc3
Genomic Map