Assay Details
| Gene Symbol: Hacd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021345.2 | NP_067320.2 | 4 - 5 | 542 | 79 | |
| GenBank mRNA | AK050556.1 | - | 4 - 5 | 536 | 79 | |
| AK051735.1 | - | 4 - 5 | 501 | 79 | ||
| AK052865.1 | - | 4 - 5 | 539 | 79 | ||
| AK159494.1 | - | 4 - 5 | 508 | 79 | ||
| BC031755.1 | - | 4 - 5 | 526 | 4488391 | 79 | |
| BC057023.1 | - | 4 - 5 | 518 | 6813640 | 79 | |
| Z97207.2 | - | 4 - 5 | 368 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hacd3
Genomic Map