Assay Details
| Gene Symbol: Gnb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029862.1 | - | 2 - 3 | 147 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thioredoxin reductase 2 | Txnrd2 |
AA118373, ESTM573010, TGR, Tr3, Trxr2, Trxrd2 |
-20.0kb | Sense | Chr. 16 - 18426417 - 18479073 on NCBI Build GRCm38 |
| Downstream | T-box 1 | Tbx1 | 80.2kb | Anti-sense | Chr. 16 - 18581704 - 18590671 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gnb1l
Genomic Map