Assay Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 15 - 16 | 1900 | 76 | |
| GenBank mRNA | AK032029.1 | - | 15 - 16 | 1901 | 76 | |
| AK080582.1 | - | 15 - 16 | 2029 | 76 | ||
| BC118010.2 | - | 14 - 15 | 1815 | 40096690 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grik4
Genomic Map