Assay Details
| Gene Symbol: Krtdap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC145437.1 | - | 2 - 3 | 156 | 9053995 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtdap| Gene Symbol: Krtdap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC145437.1 | - | 2 - 3 | 156 | 9053995 | 76 |
Set Membership: |
|