Assay Details
| Gene Symbol: Gm38431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038081.2 | 16 - 17 | 2588 | 73 | ||
| GenBank mRNA | AK004919.1 | - | 16 - 17 | 2581 | 73 | |
| AY275472.1 | - | 16 - 17 | 2567 | 73 | ||
| Gene Symbol: Fdx1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039824.2 | NP_001034913.1 | 4 - 5 | 377 | 73 | |
| GenBank mRNA | AK008401.2 | - | 4 - 5 | 375 | 73 | |
| AK010211.1 | - | 4 - 5 | 348 | 73 | ||
| AK020979.1 | - | 4 - 5 | 381 | 73 | ||
| BC061189.1 | - | 3 - 4 | 464 | 73 | ||
| BC172154.1 | - | 3 - 4 | 547 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061189.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345296 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643472 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163175 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26345303 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01881494 | - | - |
| Whole Mouse Genome G4122A | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | BC061189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | BC172154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | AK020979.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | NM_001039824.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318544 | AK008401.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08032936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08043879 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07589115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08043877 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08032937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26345261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26345252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07455280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07507361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07882164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26345269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591429 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591423 | - | - |
| Mouse Genome 430 2.0 Array | 1430293_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430293_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68165_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68165_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015578.mm.1 | NM_001039824.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015578.mm.1 | NM_001039824.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015578.mm.1 | NR_038081.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015578.mm.1 | TC0900001857.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430293_PM_a_at | - | - |
| Mouse Expression Set 430 | 1430293_a_at | - | - |
| Murine Genome U74v2 Set | 104044_at | - | - |
| Murine Genome U74v2 Set | 104044_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430293_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430293_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 386767 | - | - |
| ABI Mouse Genome Survey Microarray | 926774 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631342 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631342 | - | - |

Gm38431
Genomic Map