Assay Details
| Gene Symbol: Fxyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019503.4 | NP_062376.2 | 6 - 7 | 368 | 96 | |
| NM_052991.4 | NP_443717.1 | 6 - 7 | 351 | 96 | ||
| NM_052992.3 | NP_443718.1 | 6 - 7 | 432 | 96 | ||
| NM_194321.2 | NP_919302.1 | 6 - 7 | 415 | 96 | ||
| GenBank mRNA | AK002585.1 | - | 6 - 7 | 355 | 96 | |
| AK003504.1 | - | 2 - 3 | 150 | 96 | ||
| BC024671.1 | - | 6 - 7 | 305 | 4208962 | 96 | |
| CA459152.1 | - | 6 - 7 | 373 | 96 | ||
| CK793316.1 | - | 6 - 7 | 350 | 96 | ||
More Information
Set Membership: |
|

Fxyd1
Genomic Map