Assay Details
| Gene Symbol: Gm32511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110517.1 | 1 - 2 | 686 | 75 | ||
| GenBank mRNA | AK076745.1 | - | 1 - 2 | 688 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aquaporin 9 | Aqp9 |
1700020I22Rik, AI266899, AQP-9 |
-691.0 | Anti-sense | Chr. 9 - 71110659 - 71168085 on NCBI Build GRCm38 |
| Downstream | predicted gene 3458 | Gm3458 | 39.2kb | Anti-sense | Chr. 9 - 71211913 - 71215386 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm32511
Genomic Map