Assay Details
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213614.2 | NP_998779.2 | 5 - 6 | 456 | 62 | |
| GenBank mRNA | AF033350.1 | - | 3 - 4 | 303 | 62 | |
| AK134700.1 | - | 5 - 6 | 393 | 62 | ||
| BC059848.1 | - | 3 - 4 | 334 | 62 | ||
| BC094347.1 | - | 5 - 6 | 429 | 62 | ||
| BC141073.1 | - | 5 - 6 | 466 | 9055685 | 62 | |
| BC145331.1 | - | 5 - 6 | 466 | 9053887 | 62 | |
| BC145332.1 | - | 5 - 6 | 479 | 9053888 | 62 | |
| CA945297.1 | - | 5 - 6 | 472 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sept5
Genomic Map