Assay Details
| Gene Symbol: Sept5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213614.2 | NP_998779.2 | 7 - 8 | 700 | 92 | |
| GenBank mRNA | AF033350.1 | - | 5 - 6 | 547 | 92 | |
| AK134700.1 | - | 7 - 8 | 637 | 92 | ||
| BC059848.1 | - | 5 - 6 | 578 | 92 | ||
| BC094347.1 | - | 7 - 8 | 673 | 92 | ||
| BC141073.1 | - | 7 - 8 | 710 | 9055685 | 92 | |
| BC145331.1 | - | 7 - 8 | 710 | 9053887 | 92 | |
| BC145332.1 | - | 7 - 8 | 723 | 9053888 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sept5
Genomic Map