Assay Details
| Gene Symbol: Fam118b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286604.1 | NP_001273533.1 | 1 - 2 | 87 | 54 | |
| GenBank mRNA | AK079397.1 | - | 1 - 2 | 60 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK079397.1, NM_001286604.1 transcripts.
Set Membership: |
|

Fam118b
Genomic Map