Assay Details
| Gene Symbol: 4933433G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045851.1 | 6 - 7 | 611 | 129 | ||
| GenBank mRNA | AK017041.1 | - | 6 - 7 | 614 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphatidylserine synthase 1 | Ptdss1 |
AU044268, AW539008, PSS-1, mKIAA0024 |
-1.6kb | Anti-sense | Chr. 13 - 66932830 - 66998401 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 712 | Zfp712 |
4921504N20Rik, mszf31, mszf89 |
35.3kb | Sense | Chr. 13 - 67038594 - 67061170 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933433G19Rik
Genomic Map