Assay Details
| Gene Symbol: Gm26604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048019.1 | - | 202 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase kinase 1 | Map4k1 |
Hpk1, mHPK1 |
-68.3kb | Sense | Chr. 7 - 28982050 - 29003279 on NCBI Build GRCm38 |
| Downstream | microRNA 1963 | Mir1963 |
mir-1963, mmu-mir-1963 |
6.7kb | Anti-sense | Chr. 7 - 29083636 - 29083694 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm26604
Genomic Map