Assay Details
| Gene Symbol: Gm26787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007186.1 | - | 1 - 2 | 157 | 177 | |
| AK018988.1 | - | 1 - 2 | 134 | 177 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae) |
Stt3a |
AA408947, BB081708, Itm1 |
-236.0 | Anti-sense | Chr. 9 - 36729345 - 36767674 on NCBI Build GRCm38 |
| Downstream | etoposide induced 2.4 mRNA | Ei24 |
AA536736, AI115355, PIG8 |
2.8kb | Anti-sense | Chr. 9 - 36779153 - 36797393 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26787
Genomic Map