Assay Details
| Gene Symbol: Plet1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040714.1 | 2 - 3 | 433 | 111 | ||
| GenBank mRNA | AK009336.1 | - | 2 - 3 | 432 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | placenta expressed transcript 1 | Plet1 |
0610037B23Rik, 1600029D21Rik, AA407923, AU043048 |
Anti-sense | Chr. 9 - 50494525 - 50505639 on NCBI Build GRCm38 | |
| Downstream | 6-pyruvoyl-tetrahydropterin synthase | Pts | PTPS | 22.4kb | Sense | Chr. 9 - 50521617 - 50528641 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Plet1os
Genomic Map