Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290761.1 | NP_001277690.1 | 2 - 3 | 701 | 101 | |
| NM_001290762.1 | NP_001277691.1 | 2 - 3 | 701 | 101 | ||
| NM_001290763.1 | NP_001277692.1 | 2 - 3 | 701 | 101 | ||
| GenBank mRNA | AK012340.1 | - | 2 - 3 | 457 | 101 | |
| AK078389.1 | - | 2 - 3 | 457 | 101 | ||
| AK146521.1 | - | 456 | 101 | |||
More Information
Set Membership: |
|

Nmral1
Genomic Map