Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290761.1 | NP_001277690.1 | 3 - 4 | 955 | 82 | |
| NM_001290762.1 | NP_001277691.1 | 3 - 4 | 955 | 82 | ||
| NM_001290763.1 | NP_001277692.1 | 3 - 4 | 955 | 82 | ||
| NM_026393.2 | NP_080669.1 | 4 - 5 | 985 | 82 | ||
| GenBank mRNA | AK078389.1 | - | 3 - 4 | 711 | 82 | |
| AK146521.1 | - | 710 | 83 | |||
| AK195249.1 | - | 2 - 3 | 283 | 82 | ||
| BC030039.1 | - | 4 - 5 | 688 | 1313824 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmral1
Genomic Map