Assay Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290762.1 | NP_001277691.1 | 4 - 5 | 1138 | 68 | |
| GenBank mRNA | AK012340.1 | - | 4 - 5 | 894 | 68 | |
More Information
Set Membership: |
|

Nmral1
Genomic Map