Assay Details
| Gene Symbol: Jam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023277.4 | NP_075766.1 | 2 - 3 | 195 | 64 | |
| GenBank mRNA | AJ300304.1 | - | 2 - 3 | 155 | 64 | |
| AK003326.1 | - | 2 - 3 | 155 | 64 | ||
| AK013156.1 | - | 2 - 3 | 154 | 64 | ||
| AK032833.1 | - | 2 - 3 | 155 | 64 | ||
| AK160648.1 | - | 2 - 3 | 193 | 64 | ||
| AW106511.1 | - | 2 - 3 | 195 | 64 | ||
| BC024357.1 | - | 2 - 3 | 167 | 4973185 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jam3
Genomic Map