Assay Details
| Gene Symbol: Jam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023277.4 | NP_075766.1 | 7 - 8 | 903 | 84 | |
| GenBank mRNA | AJ300304.1 | - | 7 - 8 | 863 | 84 | |
| AK003326.1 | - | 7 - 8 | 863 | 84 | ||
| AK008187.1 | - | 7 - 8 | 896 | 84 | ||
| AK013156.1 | - | 7 - 8 | 862 | 84 | ||
| AK017692.1 | - | 111 | 84 | |||
| AK032833.1 | - | 7 - 8 | 863 | 84 | ||
| BC024357.1 | - | 7 - 8 | 875 | 4973185 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jam3
Genomic Map