Assay Details
| Gene Symbol: Ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038642.1 | NP_001033731.1 | 1 - 2 | 419 | 80 | |
| NM_011808.2 | NP_035938.2 | 1 - 2 | 419 | 80 | ||
| GenBank mRNA | AK076233.1 | - | 1 - 2 | 429 | 80 | |
| AK168247.1 | - | 1 - 2 | 376 | 80 | ||
| BB621050.1 | - | 1 - 2 | 437 | 80 | ||
| BC010588.1 | - | 1 - 2 | 414 | 3676286 | 80 | |
| BC114351.2 | - | 1 - 2 | 129 | 40056547 | 80 | |
| CD550586.1 | - | 1 - 2 | 70 | 80 | ||
| X53953.1 | - | 586 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ets1
Genomic Map