Assay Details
| Gene Symbol: Ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038642.1 | NP_001033731.1 | 3 - 4 | 749 | 75 | |
| NM_011808.2 | NP_035938.2 | 3 - 4 | 749 | 75 | ||
| GenBank mRNA | AK076233.1 | - | 3 - 4 | 759 | 75 | |
| AK168247.1 | - | 3 - 4 | 706 | 75 | ||
| AY035702.1 | - | 3 - 4 | 386 | 75 | ||
| BC010588.1 | - | 3 - 4 | 744 | 3676286 | 75 | |
| BC013089.1 | - | 2 - 3 | 266 | 75 | ||
| BC114351.2 | - | 3 - 4 | 459 | 40056547 | 75 | |
| CD550586.1 | - | 3 - 4 | 400 | 75 | ||
| M58482.1 | - | 3 - 4 | 531 | 75 | ||
| X53953.1 | - | 916 | 75 | |||
| X55787.1 | - | 3 - 4 | 497 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ets1
Genomic Map